Gene Transcripts

Gene Symbol
Pramel5
Entrez Gene ID
384077
Gene Name
preferentially expressed antigen in melanoma like 5
Gene Aliases
OTTMUSG00000010540
Chromosome Location
Chr. 4: 144270633 - 144280466 on Build GRCm38
UniGene ID
Mm.332031
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085418.2 NP_001078887.1 4 1480
GenBank mRNA BC052827.1 - 4 1490

View Full Product Details