Gene Transcripts

Gene Symbol
Prr19
Entrez Gene ID
623131
Gene Name
proline rich 19
Gene Aliases
EG623131
Chromosome Location
Chr. 7: 25301359 - 25304140 on Build GRCm38
UniGene ID
Mm.172160
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081294.1 NP_001074763.1 2 652
GenBank mRNA BC147639.1 - 2 600
BC147641.1 - 2 600 9056631
BC147701.1 - 2 600 9087460
BC147708.1 - 2 600 9087467

View Full Product Details