Gene Transcripts

Gene Symbol
Isoc2a
Entrez Gene ID
664994
Gene Name
isochorismatase domain containing 2a
Gene Aliases
Isoc2
Chromosome Location
Chr. 7: 4877053 - 4895717 on Build GRCm38
UniGene ID
Mm.302624
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101598.1 NP_001095068.1 5 609
GenBank mRNA BC158139.1 - 5 509
BG404719.1 - 4 779

View Full Product Details