Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024211.3 | NP_077173.1 | 3 | 556 | |
GenBank mRNA | AK003957.1 | - | 2 | 195 | |
AK009487.1 | - | 3 | 502 | ||
AK009824.1 | - | 3 | 456 | ||
AK168951.1 | - | 3 | 481 | ||
AK181540.1 | - | 2 | 238 | ||
AK212146.1 | - | 2 | 228 | ||
AK213879.1 | - | 2 | 238 | ||
AK215545.1 | - | 2 | 228 | ||
BC003455.1 | - | 3 | 532 | 2648336 | |
BC019631.1 | - | 3 | 510 | 4023179 |