Gene Transcripts

Gene Symbol
Hmgxb4
Entrez Gene ID
70823
Gene Name
HMG box domain containing 4
Gene Aliases
4733401K04Rik, AI316803, E430025G12, Hmgb2l1
Chromosome Location
Chr. 8: 74993575 - 75031978 on Build GRCm38
UniGene ID
Mm.261739
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178017.1 NP_821136.1 5 446
GenBank mRNA AK014640.1 - 5 466
BC023721.1 - 5 451
BC048188.1 - 6 561
BC049800.1 - 5 446 6509446

View Full Product Details