Gene Transcripts

Gene Symbol
Otud1
Entrez Gene ID
71198
Gene Name
OTU domain containing 1
Gene Aliases
4933428L19Rik
Chromosome Location
Chr. 2: 19658062 - 19660590 on Build GRCm38
UniGene ID
Mm.83981
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027715.1 NP_081991.1 1 1301
GenBank mRNA AK016972.1 - 1 608

View Full Product Details