Gene Transcripts

Gene Symbol
Tbkbp1
Entrez Gene ID
73174
Gene Name
TBK1 binding protein 1
Gene Aliases
3110043L15Rik, ProSAPiP2, SINTBAD
Chromosome Location
Chr. 11: 97136171 - 97151490 on Build GRCm38
UniGene ID
Mm.27882
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198100.2 NP_932768.2 1 193
GenBank mRNA AK014173.1 - 2 155
BC057590.1 - 1 193 6810986

View Full Product Details