Gene Transcripts

Gene Symbol
Steap2
Entrez Gene ID
74051
Gene Name
six transmembrane epithelial antigen of prostate 2
Gene Aliases
4921538B17Rik, AI930049, AW045895, IPCA-1, IPCA1, PCANAP1, STAMP1, STMP
Chromosome Location
Chr. 5: 5664829 - 5694568 on Build GRCm38
UniGene ID
Mm.274956
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001103156.2 NP_001096626.1 3, 4 788
NM_001103157.2 NP_001096627.1 3, 4 1143
NM_001285469.1 NP_001272398.1 3, 4 697
NM_001285470.1 NP_001272399.1 2, 3 662
NM_001285471.1 NP_001272400.1 2, 3 662
NM_028734.5 NP_083010.2 3, 4 845
GenBank mRNA AK052981.1 - 3, 4 847
AK086787.1 - 3, 4 815
AK089927.1 - 2, 3 636
AK145468.1 - 3, 4 734
AK162343.1 - 2, 3 624
AK165644.1 - 3, 4 1108
AK166162.1 - 3, 4 812
AK166607.1 - 3, 4 734
BC043459.1 - 3, 4 698
BC150881.1 - 3, 4 704 9087792
BC150889.1 - 3, 4 704 9087800
BC150997.1 - 755 9087910
BC151008.1 - 755 9087921

View Full Product Details