Gene Transcripts

Gene Symbol
Casc5
Entrez Gene ID
76464
Gene Name
cancer susceptibility candidate 5
Gene Aliases
2310043D08Rik, 5730505K17Rik
Chromosome Location
Chr. 2: 119047038 - 119104829 on Build GRCm38
UniGene ID
Mm.202518
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029617.2 NP_083893.2 8 2121
GenBank mRNA BC080815.1 - 8 2439

View Full Product Details