Gene Transcripts

Gene Symbol
Pabpn1
Entrez Gene ID
116697
Gene Name
poly(A) binding protein, nuclear 1
Gene Aliases
Chromosome Location
Chr. 15: 33551157 - 33560251 on Build Rnor_6.0
UniGene ID
Rn.39199
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135008.1 NP_001128480.1 5 698
XM_008770659.1 XP_008768881.1 6 2353

View Full Product Details