Gene Transcripts

Gene Symbol
Svop
Entrez Gene ID
171442
Gene Name
SV2 related protein
Gene Aliases
Chromosome Location
Chr. 12: 48292284 - 48351974 on Build Rnor_6.0
UniGene ID
Rn.162148
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134404.2 NP_599231.2 10 1097
XM_008769315.1 XP_008767537.1 10 1159
XM_008769316.1 XP_008767538.1 10 1625
GenBank mRNA AF060173.1 - 10 1254

View Full Product Details