Gene Transcripts

Gene Symbol
Jak2
Entrez Gene ID
24514
Gene Name
Janus kinase 2
Gene Aliases
Chromosome Location
Chr. 1: 247398667 - 247457521 on Build Rnor_6.0
UniGene ID
Rn.18909
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031514.1 NP_113702.1 7 812
GenBank mRNA U13396.1 - 7 812

View Full Product Details