Gene Transcripts

Gene Symbol
Fam21c
Entrez Gene ID
297530
Gene Name
family with sequence similarity 21, member C
Gene Aliases
Fam21, NP61201
Chromosome Location
Chr. 4: 148139512 - 148197361 on Build Rnor_6.0
UniGene ID
Rn.105926
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199207.1 NP_954677.1 3 311
XM_006237147.2 XP_006237209.1 3 335
XM_006237149.2 XP_006237211.1 3 335
GenBank mRNA AY257251.1 - 3 311

View Full Product Details