Gene Transcripts

Gene Symbol
Avl9
Entrez Gene ID
312371
Gene Name
AVL9 homolog (S. cerevisiase)
Gene Aliases
RGD1564615
Chromosome Location
Chr. 4: 87026476 - 87070738 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_002729314.4 XP_002729360.1 10 1291
XM_006236564.2 XP_006236626.1 10 1293
XM_006236565.2 XP_006236627.1 10 1293

View Full Product Details