Gene Transcripts

Gene Symbol
Klhl30
Entrez Gene ID
316624
Gene Name
kelch-like family member 30
Gene Aliases
RGD1309644
Chromosome Location
Chr. 9: 98490602 - 98500880 on Build Rnor_6.0
UniGene ID
Rn.21220
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750733.2 XP_003750781.1 8 1847
GenBank mRNA FQ216933.1 - 8 1815
FQ216966.1 - 8 1847

View Full Product Details