Gene Transcripts

Gene Symbol
Cmah
Entrez Gene ID
361245
Gene Name
cytidine monophospho-N-acetylneuraminic acid hydroxylase
Gene Aliases
Cmahp
Chromosome Location
Chr. 17: 42583939 - 42640221 on Build Rnor_6.0
UniGene ID
Rn.14280
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024273.1 NP_001019444.1 8, 9 1055
GenBank mRNA BC095843.1 - 8, 9 1055

View Full Product Details