Gene Transcripts

Gene Symbol
Afg3l1
Entrez Gene ID
361436
Gene Name
AFG3(ATPase family gene 3)-like 1 (S. cerevisiae)
Gene Aliases
Chromosome Location
Chr. 19: 56272171 - 56299323 on Build Rnor_6.0
UniGene ID
Rn.222173
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108456.3 NP_001101926.3 5 513
GenBank mRNA BC168848.1 - 5 406

View Full Product Details