Gene Transcripts

Gene Symbol
Myrfl
Entrez Gene ID
362879
Gene Name
myelin regulatory factor-like
Gene Aliases
RGD1562127
Chromosome Location
Chr. 7: 59799181 - 59920185 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008765434.1 XP_008763656.1 14 1717

View Full Product Details