Gene Transcripts

Gene Symbol
Upk1a
Entrez Gene ID
365227
Gene Name
uroplakin 1A
Gene Aliases
Chromosome Location
Chr. 1: 89060581 - 89071579 on Build Rnor_6.0
UniGene ID
Rn.140554
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108911.1 NP_001102381.1 4 504
XM_006228782.2 XP_006228844.1 6 861
XM_006228784.2 XP_006228846.1 6 861
XM_008759176.1 XP_008757398.1 5 841

View Full Product Details