Gene Transcripts

Gene Symbol
Cds2
Entrez Gene ID
114101
Gene Name
CDP-diacylglycerol synthase (phosphatidate cytidylyltransferase) 2
Gene Aliases
Chromosome Location
Chr. 3: 124896657 - 124935221 on Build Rnor_6.0
UniGene ID
Rn.212746
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053643.1 NP_446095.1 1, 2 134
GenBank mRNA AB052898.1 - 1, 2 134

View Full Product Details