Gene Transcripts

Gene Symbol
Klrd1
Entrez Gene ID
25110
Gene Name
killer cell lectin-like receptor, subfamily D, member 1
Gene Aliases
Cd94
Chromosome Location
Chr. 4: 163358358 - 163369925 on Build Rnor_6.0
UniGene ID
Rn.6025
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012745.2 NP_036877.1 7 904
XM_006237078.2 XP_006237140.1 7 953
GenBank mRNA AF009133.1 - 7 661
BC086393.1 - 7 904 7303354

View Full Product Details