Gene Transcripts

Gene Symbol
Ccl6
Entrez Gene ID
287910
Gene Name
chemokine (C-C motif) ligand 6
Gene Aliases
Mrp-1, Scay6
Chromosome Location
Chr. 10: 70798118 - 70802750 on Build Rnor_6.0
UniGene ID
Rn.7857
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004202.2 NP_001004202.1 1 21
GenBank mRNA AY620903.1 - 1 31
BC079460.1 - 53 7121977
EF121992.1 - 1 21
EF121993.1 - 1 21

View Full Product Details