Gene Transcripts

Gene Symbol
Nkx2-5
Entrez Gene ID
114109
Gene Name
NK2 homeobox 5
Gene Aliases
Csx, Nkx2.5
Chromosome Location
Chr. 10: 16635989 - 16638758 on Build Rnor_6.0
UniGene ID
Rn.6179
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053651.1 NP_446103.1 2 821
GenBank mRNA AF006664.1 - 2 821

View Full Product Details