Gene Transcripts

Gene Symbol
Abcc2
Entrez Gene ID
25303
Gene Name
ATP-binding cassette, subfamily C (CFTR/MRP), member 2
Gene Aliases
Cmoat, Mrp2, cMRP
Chromosome Location
Chr. 1: 263554545 - 263612541 on Build Rnor_6.0
UniGene ID
Rn.10265
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012833.2 NP_036965.1 28 4140
GenBank mRNA D86086.1 - 28 3962
L49379.1 - 28 4021
X96393.1 - 28 3976

View Full Product Details