Gene Transcripts

Gene Symbol
Atf3
Entrez Gene ID
25389
Gene Name
activating transcription factor 3
Gene Aliases
LRF-1, LRFI
Chromosome Location
Chr. 13: 109817892 - 109849632 on Build Rnor_6.0
UniGene ID
Rn.9664
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012912.2 NP_037044.1 2 297
GenBank mRNA BC078903.1 - 2 297
M63282.1 - 2 302

View Full Product Details