Gene Transcripts

Gene Symbol
Med10
Entrez Gene ID
290939
Gene Name
mediator complex subunit 10
Gene Aliases
RGD1310128
Chromosome Location
Chr. 1: 36107249 - 36118883 on Build Rnor_6.0
UniGene ID
Rn.1773
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106097.2 NP_001099567.2 2, 3 258
XM_006227769.2 XP_006227831.1 3, 4 574
GenBank mRNA CB582092.1 - 2, 3 258

View Full Product Details