Gene Transcripts

Gene Symbol
Cfb
Entrez Gene ID
294257
Gene Name
complement factor B
Gene Aliases
Bf, Da1-24
Chromosome Location
Chr. 20: 4536206 - 4542073 on Build Rnor_6.0
UniGene ID
Rn.109148
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_212466.3 NP_997631.2 1 240
GenBank mRNA BC087089.1 - 1 240

View Full Product Details