Gene Transcripts

Gene Symbol
Malsu1
Entrez Gene ID
297082
Gene Name
mitochondrial assembly of ribosomal large subunit 1
Gene Aliases
RGD1306936
Chromosome Location
Chr. 4: 78735279 - 78744102 on Build Rnor_6.0
UniGene ID
Rn.35387
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106593.1 NP_001100063.1 4 672

View Full Product Details