Gene Transcripts

Gene Symbol
Dnajc27
Entrez Gene ID
298859
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 27
Gene Aliases
Rbj
Chromosome Location
Chr. 6: 28515076 - 28544758 on Build Rnor_6.0
UniGene ID
Rn.22692
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_206845.2 NP_996727.2 5 641
GenBank mRNA AY539931.1 - 7 783
DY314277.1 - 5 641

View Full Product Details