Gene Transcripts

Gene Symbol
Dhrs7
Entrez Gene ID
299131
Gene Name
dehydrogenase/reductase (SDR family) member 7
Gene Aliases
DR-NR#3
Chromosome Location
Chr. 6: 95480042 - 95502845 on Build Rnor_6.0
UniGene ID
Rn.119024
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013098.1 NP_001013116.1 5 814
GenBank mRNA AB108671.1 - 5 814
BC104713.1 - 5 720 7370181

View Full Product Details