Gene Transcripts

Gene Symbol
Lrrc47
Entrez Gene ID
362672
Gene Name
leucine rich repeat containing 47
Gene Aliases
RGD1308599
Chromosome Location
Chr. 5: 171297850 - 171307682 on Build Rnor_6.0
UniGene ID
Rn.160689
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135666.1 NP_001129138.1 5 1371

View Full Product Details