Gene Transcripts

Gene Symbol
LOC367195
Entrez Gene ID
367195
Gene Name
similar to 60S RIBOSOMAL PROTEIN L7
Gene Aliases
Ab2-073
Chromosome Location
Chr. 8: 132026578 - 132029465 on Build Rnor_6.0
UniGene ID
Rn.202516
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047920.1 NP_001041385.1 2 401
GenBank mRNA AY325186.1 - 2 401

View Full Product Details