Gene Transcripts

Gene Symbol
Stpg2
Entrez Gene ID
499719
Gene Name
sperm-tail PG-rich repeat containing 2
Gene Aliases
Chromosome Location
Chr. 2: 244521605 - 245044919 on Build Rnor_6.0
UniGene ID
Rn.215031
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100993.1 NP_001094463.1 11 1489
XM_008761520.1 XP_008759742.1 11 1583
GenBank mRNA BC128777.1 - 11 1489

View Full Product Details