Gene Transcripts

Gene Symbol
Lace1
Entrez Gene ID
502479
Gene Name
lactation elevated 1
Gene Aliases
Chromosome Location
Chr. 20: 47038824 - 47208812 on Build Rnor_6.0
UniGene ID
Rn.24314
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037656.1 NP_001032745.1 8 1260
GenBank mRNA BC107937.1 - 8 1260

View Full Product Details