Gene Transcripts

Gene Symbol
Crh
Entrez Gene ID
81648
Gene Name
corticotropin releasing hormone
Gene Aliases
CRF
Chromosome Location
Chr. 2: 104459999 - 104461863 on Build Rnor_6.0
UniGene ID
Rn.10349
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031019.1 NP_112281.1 2 722
GenBank mRNA X03036.1 - 2 720

View Full Product Details