Gene Transcripts

Gene Symbol
Spag4
Entrez Gene ID
83623
Gene Name
sperm associated antigen 4
Gene Aliases
Chromosome Location
Chr. 3: 151609602 - 151613946 on Build Rnor_6.0
UniGene ID
Rn.28620
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031792.1 NP_113980.1 9 955
XM_008762386.1 XP_008760608.1 9 689
GenBank mRNA AF043345.2 - 9 955

Gene Symbol
LOC100911109
Entrez Gene ID
100911109
Gene Name
sperm-associated antigen 4 protein-like
Gene Aliases
Chromosome Location
Chr. 3: 152143565 - 152148051 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749611.3 XP_003749659.1 1097
XM_008762411.1 XP_008760633.1 689

View Full Product Details