Gene Transcripts

Gene Symbol
Dnase1l3
Entrez Gene ID
13421
Gene Name
deoxyribonuclease 1-like 3
Gene Aliases
DNasegamma, Dhp2, Lsd
Chromosome Location
Chr.14: 7965187 - 7994653 on Build GRCm38
Uni Gene
Mm.272258
Interrogated Sequence Translated Protein
RefSeq NM_007870.3 NP_031896.3
GenBank mRNA AF047355.1 -
AK156109.1 -
AK164072.1 -
AK171843.1 -
AK172254.1 -
BC012671.1 -
U76110.1 -

View Full Product Details