Gene Transcripts

Gene Symbol
Map4k3
Entrez Gene ID
170920
Gene Name
mitogen-activated protein kinase kinase kinase kinase 3
Gene Aliases
-
Chromosome Location
Chr.6: 3276732 - 3444643 on Build Rnor_6.0
Uni Gene
Rn.93345
Interrogated Sequence Translated Protein
RefSeq NM_133407.1 NP_596898.1
XM_006239656.2 XP_006239718.1
GenBank mRNA AA819812.1 -
AF312224.1 -
AI502900.1 -
CK474868.1 -
DV716943.1 -

View Full Product Details