Gene Transcripts

Gene Symbol
Y37F4.3
Entrez Gene ID
260019
Gene Name
Uncharacterized protein
Gene Aliases
CELE_Y37F4.3
Chromosome Location
Chr.I: 10850405 - 10851268 on Build WBcel235
Uni Gene
Cel.25431
Interrogated Sequence Translated Protein
RefSeq NM_170920.3 NP_740915.1

View Full Product Details