Gene Transcripts

Gene Symbol
Phlpp
Entrez Gene ID
35178
Gene Name
PH domain leucine-rich repeat protein phosphatase
Gene Aliases
CG10493, CT29458, Dmel\CG10493, Dmel_CG10493, PP2C, dPHLPP
Chromosome Location
Chr.2L: 19059129 - 19063132 on Build Release_6_plus_ISO1_MT
Uni Gene
Dm.17963
Species Specific ID (Flybase ID)
FBgn0032749
Interrogated Sequence Translated Protein
RefSeq NM_001273629.1 NP_001260558.1
NM_136094.2 NP_609938.1

View Full Product Details