Gene Transcripts
- Gene Symbol
- Phlpp
- Entrez Gene ID
-
35178
- Gene Name
- PH domain leucine-rich repeat protein phosphatase
- Gene Aliases
-
CG10493, CT29458, Dmel\CG10493, Dmel_CG10493, PP2C, dPHLPP
- Chromosome Location
-
Chr.2L: 19059129 - 19063132 on Build Release_6_plus_ISO1_MT
- Uni Gene
- Dm.17963
- Species Specific ID (Flybase ID)
- FBgn0032749
View Full Product Details