Gene Transcripts

Gene Symbol
Vrp1
Entrez Gene ID
37521
Gene Name
Verprolin 1
Gene Aliases
CG13503, D-WIP, DWIP, DWip, Dm_Vrp1, Dmel\CG13503, Dmel_CG13503, Sltr, WIP, Wip, dWIP, sltr, solas, vrp, vrp1, wip
Chromosome Location
Chr.2R: 22106107 - 22122562 on Build Release_6_plus_ISO1_MT
Uni Gene
Dm.7264
Species Specific ID (Flybase ID)
FBgn0243516
Interrogated Sequence Translated Protein
RefSeq NM_001103936.2 NP_001097406.1
NM_001169775.2 NP_001163246.1
NM_001169776.2 NP_001163247.1
NM_001169777.2 NP_001163248.1
NM_001169778.2 NP_001163249.1
NM_001299775.1 NP_001286704.1
NM_137796.3 NP_611640.3
NM_166507.2 NP_726159.1
NM_166508.2 NP_726160.1
NM_166509.2 NP_726161.1
NM_166510.2 NP_726162.1
NM_166511.2 NP_726163.1
GenBank mRNA AY094731.1 -

View Full Product Details