Gene Transcripts

Gene Symbol
Snhg6
Entrez Gene ID
73824
Gene Name
small nucleolar RNA host gene 6
Gene Aliases
1110008H02Rik
Chromosome Location
Chr. 1: 9942025 - 9944118 on Build GRCm38
UniGene ID
Mm.28311
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024067.2 3, 4 268
GenBank mRNA AK003573.1 - 3, 4 269


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CJ320246.1 - 2 135 Chr. 1 - 9942026 - 9942683 [-]
GenBankmRNA CJ082230.1 - 3, 4 267 Chr. 1 - 9942037 - 9944117 [-]
GenBankmRNA AA015539.1 - 3, 4 228 Chr. 1 - 9942042 - 9944079 [-]
GenBankmRNA AA023387.1 - 3, 4 230 Chr. 1 - 9942023 - 9944083 [-]
GenBankmRNA BY604390.1 - 3, 4 273 Chr. 1 - 9942019 - 9944118 [-]
GenBankmRNA BE853719.1 - 3 253 Chr. 1 - 9942153 - 9944103 [-]
GenBankmRNA AV115890.2 - 447 Chr. - - [-]
GenBankmRNA AA250387.1 - 3, 4 245 Chr. 1 - 9942054 - 9944087 [-]
GenBankmRNA BY345355.1 - 3, 4 271 Chr. 1 - 9942107 - 9944121 [-]
GenBankmRNA BY111725.1 - 3, 4 267 Chr. 1 - 9942091 - 9944117 [-]
GenBankmRNA AV485517.1 - 3, 4 255 Chr. 1 - 9942023 - 9944104 [-]
GenBankmRNA BI109518.1 - 3, 4 251 Chr. 1 - 9942023 - 9944099 [-]
GenBankmRNA AV468093.1 - 3, 4 228 Chr. 1 - 9942028 - 9944077 [-]
GenBankmRNA AA097989.1 - 3, 4 238 Chr. 1 - 9942025 - 9944095 [-]
GenBankmRNA BE134757.1 - 3, 4 248 Chr. 1 - 9942023 - 9944098 [-]
GenBankmRNA AA267148.1 - 2, 3 106 Chr. 1 - 9942023 - 9942654 [-]
GenBankmRNA BX523010.1 - 3, 4 258 Chr. 1 - 9942023 - 9944107 [-]
GenBankmRNA W18398.1 - 237 Chr. - - [-]
GenBankmRNA W35693.1 - 197 Chr. - - [-]
GenBankmRNA AA023086.1 - 3, 4 257 Chr. 1 - 9942024 - 9944105 [-]
GenBankmRNA AA119805.1 - 3, 4 243 Chr. 1 - 9942023 - 9944098 [-]
GenBankmRNA W40695.1 - 196 Chr. - - [-]
GenBankmRNA AA940467.1 - 3, 4 247 Chr. 1 - 9941959 - 9944090 [-]
GenBankmRNA BX516798.1 - 3, 4 258 Chr. 1 - 9942023 - 9944105 [-]
GenBankmRNA AL034794.1 - 2 358 Chr. 1 - 9942230 - 9942897 [-]
GenBankmRNA AA002823.1 - 3, 4 251 Chr. 1 - 9942023 - 9944098 [-]
GenBankmRNA AA289428.1 - 2, 3 153 Chr. 1 - 9942025 - 9942695 [-]
GenBankmRNA W54219.1 - 193 Chr. - - [-]
GenBankmRNA BY345466.1 - 3, 4 271 Chr. 1 - 9942116 - 9944121 [-]
GenBankmRNA BY243424.1 - 3, 4 268 Chr. 1 - 9942019 - 9944117 [-]
GenBankmRNA W11261.1 - 251 Chr. - - [-]
GenBankmRNA AA146116.1 - 3, 4 188 Chr. 1 - 9942033 - 9944038 [-]
GenBankmRNA AW988981.1 - 3, 4 248 Chr. 1 - 9942023 - 9944098 [-]
GenBankmRNA AA098159.1 - 3, 4 245 Chr. 1 - 9942037 - 9944097 [-]
GenBankmRNA AA014503.1 - 3 209 Chr. 1 - 9942168 - 9944058 [-]
GenBankmRNA AA155393.1 - 3, 4 193 Chr. 1 - 9942025 - 9944043 [-]
GenBankmRNA BY703829.1 - 3, 4 267 Chr. 1 - 9942025 - 9944117 [-]
GenBankmRNA BF715116.1 - 3, 4 255 Chr. 1 - 9942023 - 9944105 [-]
GenBankmRNA AA124208.1 - 3, 4 252 Chr. 1 - 9942058 - 9944104 [-]
GenBankmRNA AA240411.1 - 2, 3 106 Chr. 1 - 9942039 - 9942654 [-]
GenBankmRNA BY711012.1 - 3, 4 267 Chr. 1 - 9941960 - 9944117 [-]
GenBankmRNA W54183.1 - 250 Chr. - - [-]
GenBankmRNA AV490092.1 - 3, 4 232 Chr. 1 - 9942038 - 9944081 [-]
GenBankmRNA AV115922.2 - 3, 4 289 Chr. 1 - 9941973 - 9944118 [-]

View Full Product Details