Gene Transcripts

Gene Symbol
5430405H02Rik
Entrez Gene ID
74487
Gene Name
RIKEN cDNA 5430405H02 gene
Gene Aliases
2610307N17Rik
Chromosome Location
Chr. 2: 156852403 - 156862945 on Build GRCm38
UniGene ID
Mm.490465
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015591.1 2 1016
NR_027914.1 2 730
GenBank mRNA AK134903.1 - 2 732
AK134962.1 - 2 966
BC042697.1 - 2 837

View Full Product Details