Gene Transcripts
- Gene Symbol
- Gm10190
- Entrez Gene ID
- 791338
- Gene Name
- predicted gene 10190
- Gene Aliases
- ENSMUSG00000066938
- Chromosome Location
- Chr. 17: 80371857 - 80373542 on Build GRCm38
- UniGene ID
- Mm.204722
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028385.1 |
|
1 |
1307 |
|
GenBank mRNA |
AK034626.1 |
- |
1 |
1308 |
|
View Full Product Details