Gene Transcripts

Gene Symbol
Psg-ps1
Entrez Gene ID
232919
Gene Name
pregnancy specific glycoprotein pseudogene 1
Gene Aliases
AV036018, Cea-6, Cea6
Chromosome Location
Chr. 7: 17672313 - 17682060 on Build GRCm38
UniGene ID
Mm.153893
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002857.1 2 499
GenBank mRNA AK132281.1 - 2 499
M83348.1 - 2 495


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BY713665.1 - 2 499 Chr. 7 - 17672313 - 17677731 [+]
GenBankmRNA CK020238.1 - 2 479 Chr. 7 - 17672333 - 17677893 [+]
GenBankmRNA CK021640.1 - 2 420 Chr. 7 - 17672393 - 17677953 [+]
GenBankmRNA CK021064.1 - 2 451 Chr. 7 - 17672361 - 17677928 [+]
GenBankmRNA CK021969.1 - 2 485 Chr. 7 - 17672327 - 17677852 [+]
GenBankmRNA CK023168.1 - 2 471 Chr. 7 - 17672341 - 17677881 [+]
GenBankmRNA CK023083.1 - 2 546 Chr. 7 - 17672266 - 17677813 [+]
GenBankmRNA CF551674.1 - 2 549 Chr. 7 - 17672292 - 17678005 [+]
GenBankmRNA CK022583.1 - 2 492 Chr. 7 - 17672320 - 17677904 [+]

View Full Product Details