Gene Transcripts
- Gene Symbol
- 5430416O09Rik
- Entrez Gene ID
- 71406
- Gene Name
- RIKEN cDNA 5430416O09 gene
- Gene Aliases
- AI481365
- Chromosome Location
- Chr. 4: 43730034 - 43734534 on Build GRCm38
- UniGene ID
- Mm.33063
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033355.1 |
|
2 |
3261 |
|
GenBank mRNA |
AK030334.1 |
- |
2 |
3262 |
|
View Full Product Details