Gene Transcripts
- Gene Symbol
- TPM3P9
- Entrez Gene ID
- 147804
- Gene Name
- tropomyosin 3 pseudogene 9
- Gene Aliases
- Chromosome Location
- Chr. 19: 53431974 - 53444672 on Build GRCh38
- UniGene ID
- Hs.433293
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003148.3 |
|
2 |
1680 |
|
GenBank mRNA |
AK094777.1 |
- |
4 |
1619 |
|
BX647962.1 |
- |
2 |
1666 |
|
View Full Product Details