Gene Transcripts
- Gene Symbol
- MIR9-3HG
- Entrez Gene ID
- 254559
- Gene Name
- MIR9-3 host gene
- Gene Aliases
- LINC00925
- Chromosome Location
- Chr. 15: 89361579 - 89398487 on Build GRCh38
- UniGene ID
- Hs.136313
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DA755484.1 |
- |
3 |
455 |
|
Chr. 15 - 89362474 - 89379213 [+] |
GenBankmRNA |
DA755518.1 |
- |
3 |
455 |
|
Chr. 15 - 89362474 - 89379202 [+] |
GenBankmRNA |
DA755631.1 |
- |
3 |
455 |
|
Chr. 15 - 89362474 - 89379229 [+] |
GenBankmRNA |
DA755665.1 |
- |
3 |
456 |
|
Chr. 15 - 89362474 - 89379232 [+] |
View Full Product Details