Gene Transcripts

Gene Symbol
TIMM8B
Entrez Gene ID
26521
Gene Name
translocase of inner mitochondrial membrane 8 homolog B
Gene Aliases
DDP2, TIM8B
Chromosome Location
Chr. 11: 112084800 - 112086798 on Build GRCh38
UniGene ID
Hs.279915
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028383.1 2 353
GenBank mRNA AF165967.1 - 2 263
BC000711.2 - 2 300 2823930

View Full Product Details